Learn More
Alkali Scientific CellPro™ Carbenicillin Solution 50mg/mL, 10mL
SDP
Catalog No. 502388696
Encompass_Preferred
CellPro™ Carbenicillin is a specialized culture medium containing carbenicillin, a β-lactam antibiotic used to select for bacterial strains carrying carbenicillin resistance genes. This LB agar variant is commonly employed in molecular biology research for the cultivation and maintenance of recombinant bacterial cells harboring carbenicillin resistance plasmids. By incorporating carbenicillin into the LB agar, this medium enables the selective growth of transformed bacterial strains while inhibiting the growth of non-resistant contaminants.
Catalog No. 50-238-8696 Supplier Alkali Scientific Supplier No. DS0171
CellPro™ Carbenicillin is a specialized culture medium containing carbenicillin, a β-lactam antibiotic used to select for bacterial strains carrying carbenicillin resistance genes. This LB agar variant is commonly employed in molecular biology research for the cultivation and maintenance of recombinant bacterial cells harboring carbenicillin resistance plasmids. By incorporating carbenicillin into the LB agar, this medium enables the selective growth of transformed bacterial strains while inhibiting the growth of non-resistant contaminants.