Learn More
Alkali Scientific CellPro™ Cell Culture (TC) Grade Water, 500ml, 10/Case
SDP
Catalog No. 503027535
Encompass_Preferred
Eliminate endotoxin concerns with endotoxin-free CellPro™ Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes. Undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP). Meets < 0.005 EU/mL endotoxin by turbidimetric assay. Manufactured in our ISO 9001 certified facilities. Manufactured using ISO 9001 and ISO 13485 certified processes. CellPro™ Cell Culture Grade Water can be used in multiple cell culture applications. With endotoxin levels as low as < 0.005 EU/mL, CellPro™ Cell Culture Grade Water supports healthy cell growth and consistent experimental results.
Catalog No. 50-302-7535 Supplier Alkali Scientific Supplier No. DTC05CS
Eliminate endotoxin concerns with endotoxin-free CellPro™ Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes. Undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP). Meets < 0.005 EU/mL endotoxin by turbidimetric assay. Manufactured in our ISO 9001 certified facilities. Manufactured using ISO 9001 and ISO 13485 certified processes. CellPro™ Cell Culture Grade Water can be used in multiple cell culture applications. With endotoxin levels as low as < 0.005 EU/mL, CellPro™ Cell Culture Grade Water supports healthy cell growth and consistent experimental results.