Learn More
Alkali Scientific CellPro™ Conical Centrifuge Tubes - Sterile, 500mL, 40/Case
SDP
Catalog No. 503026591
Encompass_Preferred
The 250mL and 500mL conical centrifuge tube can process a large number of samples at one time and are suitable for separation and harvest of large cells, supernatants, bacteria, yeast and tissue samples, therefore they are widely used in the field of life sciences and clinical experiments and can meet the requirements of biological analysis.   Product Features● The unique sealing structure design of the cover provides a two-way security to guarantee seal ability.● The thickened design of the conical bottom increases the maximum centrifugal limit.● Max. centrifugal limit 7000xg.● Tolerable temperature -80℃-120℃.● The clear scale is convenient for data reading.● No pyrogens, sterile, no DNARNA.● Electron beam sterilization, SAL=10-6
Catalog No. 50-302-6591 Supplier Alkali Scientific Supplier No. C8500
The 250mL and 500mL conical centrifuge tube can process a large number of samples at one time and are suitable for separation and harvest of large cells, supernatants, bacteria, yeast and tissue samples, therefore they are widely used in the field of life sciences and clinical experiments and can meet the requirements of biological analysis.   Product Features● The unique sealing structure design of the cover provides a two-way security to guarantee seal ability.● The thickened design of the conical bottom increases the maximum centrifugal limit.● Max. centrifugal limit 7000xg.● Tolerable temperature -80℃-120℃.● The clear scale is convenient for data reading.● No pyrogens, sterile, no DNARNA.● Electron beam sterilization, SAL=10-6