Learn More
Alkali Scientific CellPro™ Cryovial Cap Inserts, for Cryogenic Vial, White, 100/pk, 1000/cs, Green
SDP
Catalog No. 5000662558
Encompass_Preferred
The CellPro™ Cryovial Cap Inserts are designed to provide secure and efficient identification for cryogenic sample vials used in laboratory storage systems. These inserts fit standard cryovial caps and allow laboratories to implement color coded sample organization for improved tracking and inventory management. Manufactured from durable cryo compatible materials, the inserts maintain stability and structural integrity at ultra low temperatures used in cryogenic storage. The color coded design helps researchers quickly identify stored samples, reducing handling errors and improving workflow efficiency. Supplied in packs of 100 with 10 packs per case, these inserts are ideal for high throughput laboratories, biorepositories, and research facilities requiring reliable sample organization. CellPro™ cap inserts support efficient cryogenic storage, biological research, and long term specimen preservation.
Catalog No. 50-006-62558 Supplier Alkali Scientific Supplier No. VCACNG
The CellPro™ Cryovial Cap Inserts are designed to provide secure and efficient identification for cryogenic sample vials used in laboratory storage systems. These inserts fit standard cryovial caps and allow laboratories to implement color coded sample organization for improved tracking and inventory management. Manufactured from durable cryo compatible materials, the inserts maintain stability and structural integrity at ultra low temperatures used in cryogenic storage. The color coded design helps researchers quickly identify stored samples, reducing handling errors and improving workflow efficiency. Supplied in packs of 100 with 10 packs per case, these inserts are ideal for high throughput laboratories, biorepositories, and research facilities requiring reliable sample organization. CellPro™ cap inserts support efficient cryogenic storage, biological research, and long term specimen preservation.