Learn More
Alkali Scientific CellPro™ Cryovial Cap Inserts, for Cryogenic Vial, White, 100/pk, 1000/cs, Yellow
SDP
Catalog No. 5000662575
Encompass_Preferred
The CellPro™ Cryovial Cap Inserts provide secure and reliable identification for cryogenic vials used in sample storage, transport, and long term preservation. Designed to fit standard cryogenic vial caps, these inserts enable clear labeling and efficient color coding of samples for improved organization in laboratories and biorepositories. Manufactured from durable, cryo compatible materials, they maintain structural integrity at ultra low storage temperatures commonly used for biological samples. The smooth surface supports easy marking or labeling for quick sample recognition. Supplied in convenient packs of 100 with 10 packs per case (1000 total), these inserts are ideal for high throughput laboratories, research facilities, clinical storage environments, and biobanking applications requiring reliable sample identification and efficient inventory management.
Catalog No. 50-006-62575 Supplier Alkali Scientific Supplier No. VCACNY
The CellPro™ Cryovial Cap Inserts provide secure and reliable identification for cryogenic vials used in sample storage, transport, and long term preservation. Designed to fit standard cryogenic vial caps, these inserts enable clear labeling and efficient color coding of samples for improved organization in laboratories and biorepositories. Manufactured from durable, cryo compatible materials, they maintain structural integrity at ultra low storage temperatures commonly used for biological samples. The smooth surface supports easy marking or labeling for quick sample recognition. Supplied in convenient packs of 100 with 10 packs per case (1000 total), these inserts are ideal for high throughput laboratories, research facilities, clinical storage environments, and biobanking applications requiring reliable sample identification and efficient inventory management.