Learn More
Alkali Scientific CellPro™ Culture Tube Cap, two position for 17×100mm tubes, PE, 8 bags of 125
SDP
Catalog No. 503029359
Encompass_Preferred
Culture tubes are now available without caps or with the caps packed separately and available in both 12×75mm (5mL) or 17×100mm (14mL) capacities. Polypropylene (PP) or Polystyrene (PS).
Catalog No. 50-302-9359 Supplier Alkali Scientific Supplier No. CT8239
Culture tubes are now available without caps or with the caps packed separately and available in both 12×75mm (5mL) or 17×100mm (14mL) capacities. Polypropylene (PP) or Polystyrene (PS).