Learn More
Alkali Scientific CellPro™ Deep Well Plate, 384 Square Well, 240µL, 24.9mm height, V-bottom, PP, 5 bags of 10 pieces, 50/Cs, STERILE
SDP
Catalog No. 503028802
Encompass_Preferred
CellPro™ plates for HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation, and all are certified non-pyrogenic, RNase/DNase free. Black printed alphanumeric coatings Over 30 plate configurations Sterile or nonsterile Private label available - no minimum U (round), V (conical) or F (flat) bottoms
Catalog No. 50-302-8802 Supplier Alkali Scientific Supplier No. DWP3843S
CellPro™ plates for HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation, and all are certified non-pyrogenic, RNase/DNase free. Black printed alphanumeric coatings Over 30 plate configurations Sterile or nonsterile Private label available - no minimum U (round), V (conical) or F (flat) bottoms