Learn More
Alkali Scientific CellPro™ Denhardt's Solution, 50X, 50mL, Frozen
SDP
Catalog No. 502389408
Encompass_Preferred
Alkali Scientific™ presents CellPro™ Denhardt's Solution, a sterile and frozen 50X concentrate solution meticulously filtered for purity. With a volume of 50mL, this solution offers convenience and efficiency for molecular biology applications. Denhardt's Solution, at a concentration of 50X, provides a highly concentrated mixture of blocking agents, including Ficoll, polyvinylpyrrolidone (PVP), and bovine serum albumin (BSA), to effectively minimize non-specific binding in nucleic acid hybridization and protein blotting assays. Its sterile filtration ensures the elimination of contaminants, maintaining the integrity of your experimental results. Ideal for various molecular biology techniques, including Southern and Northern blotting, in situ hybridization, and antibody-based detection methods. Trust in the reliability and quality of CellPro™ Denhardt's Solution for consistent and accurate outcomes in your research endeavors.
Catalog No. 50-238-9408 Supplier Alkali Scientific Supplier No. DB0247
Add to Cart
Add to Cart
Alkali Scientific™ presents CellPro™ Denhardt's Solution, a sterile and frozen 50X concentrate solution meticulously filtered for purity. With a volume of 50mL, this solution offers convenience and efficiency for molecular biology applications. Denhardt's Solution, at a concentration of 50X, provides a highly concentrated mixture of blocking agents, including Ficoll, polyvinylpyrrolidone (PVP), and bovine serum albumin (BSA), to effectively minimize non-specific binding in nucleic acid hybridization and protein blotting assays. Its sterile filtration ensures the elimination of contaminants, maintaining the integrity of your experimental results. Ideal for various molecular biology techniques, including Southern and Northern blotting, in situ hybridization, and antibody-based detection methods. Trust in the reliability and quality of CellPro™ Denhardt's Solution for consistent and accurate outcomes in your research endeavors.