Learn More
Alkali Scientific CellPro™ DEPC treated Ultra Pure Water, 1L, 12/cs
SDP
Catalog No. 502389417
Encompass_Preferred
CellPro™ DEPC treated Ultra Pure Water is specially treated with diethyl pyrocarbonate (DEPC) to inactivate ribonucleases (RNases), ensuring the purity of the water for molecular biology applications. DEPC treatment effectively eliminates RNase contamination, making this ultra-pure water ideal for use in RNA-related experiments, such as RNA isolation, reverse transcription, and RNA sequencing.
Catalog No. 50-238-9417 Supplier Alkali Scientific Supplier No. DC42236CS
CellPro™ DEPC treated Ultra Pure Water is specially treated with diethyl pyrocarbonate (DEPC) to inactivate ribonucleases (RNases), ensuring the purity of the water for molecular biology applications. DEPC treatment effectively eliminates RNase contamination, making this ultra-pure water ideal for use in RNA-related experiments, such as RNA isolation, reverse transcription, and RNA sequencing.