Learn More
Alkali Scientific CellPro™ Dialyzed Fetal Bovine Serum (FBS), US Origin, 500mL
SDP
Catalog No. 503027452
Encompass_Preferred
Alkali Scientific™ Dialyzed FBS is manufactured from US sourced FBS using a proprietary process to reduce concentrations of small molecules such as cytokines, nucleotides, amino acids, hormones, small proteins, and salts. This creates a small molecule environment which provides greater definition and control. Dialyzed FBS is ideal for applications where normal FBS small molecule concentrations may be disruptive.. It meets the highest industry standards for endotoxin and hemoglobin levels and supports the growth of the widest variety of cell lines. Triple 0.1µm sterile-filtered Mycoplasma-tested and virus-screened Ships on dry ice Store frozen at -20 to -10°C
Catalog No. 50-302-7452 Supplier Alkali Scientific Supplier No. FB108
Alkali Scientific™ Dialyzed FBS is manufactured from US sourced FBS using a proprietary process to reduce concentrations of small molecules such as cytokines, nucleotides, amino acids, hormones, small proteins, and salts. This creates a small molecule environment which provides greater definition and control. Dialyzed FBS is ideal for applications where normal FBS small molecule concentrations may be disruptive.. It meets the highest industry standards for endotoxin and hemoglobin levels and supports the growth of the widest variety of cell lines. Triple 0.1µm sterile-filtered Mycoplasma-tested and virus-screened Ships on dry ice Store frozen at -20 to -10°C