Learn More
Alkali Scientific CellPro™ Disposable Test Tubes, PP, 12 × 75mm, 5ml, 5000/Case
SDP
Catalog No. 503029172
Encompass_Preferred
Disposable Test Tubes are packaged for high volume users in bulk quantity economy cases. Supplied non-sterile and without caps (available separately), all three sizes are available in translucent polypropylene or crystal clear polystyrene. Plug-type caps are available separately. Round Bottoms Non-Sterile Plug Caps Available Separately Resealable Zip Bags Non-Graduated
Catalog No. 50-302-9172 Supplier Alkali Scientific Supplier No. CT2014
Disposable Test Tubes are packaged for high volume users in bulk quantity economy cases. Supplied non-sterile and without caps (available separately), all three sizes are available in translucent polypropylene or crystal clear polystyrene. Plug-type caps are available separately. Round Bottoms Non-Sterile Plug Caps Available Separately Resealable Zip Bags Non-Graduated