Learn More
Alkali Scientific CellPro™ DMEM, 1X, High Glucose, without L-glutamine, with Sodium Pyruvate, 100mL
SDP
Catalog No. 502389442
Encompass_Preferred
Alkali Scientific™ CellPro™ DMEM, 1x is a standard growth medium formulated for the culture of mammalian cells in vitro. With its balanced composition of amino acids, vitamins, and high glucose concentration, this media supports robust cell proliferation and metabolism. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for cell culture, cell-based assays, and biomedical research.
Catalog No. 50-238-9442 Supplier Alkali Scientific Supplier No. DMEM11
Add to Cart
Add to Cart
Alkali Scientific™ CellPro™ DMEM, 1x is a standard growth medium formulated for the culture of mammalian cells in vitro. With its balanced composition of amino acids, vitamins, and high glucose concentration, this media supports robust cell proliferation and metabolism. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for cell culture, cell-based assays, and biomedical research.