Learn More
Alkali Scientific CellPro™ DMEM/F12 1:1 medium with L-glutamine, HEPES, 500ml
SDP
Catalog No. 503027568
Encompass_Preferred
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 50-302-7568 Supplier Alkali Scientific Supplier No. F12001
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.