Learn More
Alkali Scientific CellPro DMEM/High glucose with
SDP
Catalog No. 503027118
Encompass_Preferred
DMEM high glucose liquid media supports cell growth in research upstream process development and cell culture manufacturing processes DMEM high glucose medium efficacy and homogeneity is tested DMEM media contain increased concentrations of vitamins minerals and amino acids compared to traditional basal media DMEM medium composition also includes ferric nitrate making this cell culture medium an option for certain cell lines requiring iron for cell growth DMEM media preparations are available in high or low glucose formulations as well as with or without L-glutamine magnesium and sodium pyruvate DMEM formulation options may also include phenol red as a pH indicator All available DMEM media products contain a sodium bicarbonate buffer with or without HEPES to maintain optimal culture pH DMEM raw material components are screened through strict quality control testing for lot-to-lot consistency DMEM liquid media products are hydrated using purified water and undergo sterile filtrat
Catalog No. 50-302-7118 Supplier Alkali Scientific Supplier No. DMEM04CS
DMEM high glucose liquid media supports cell growth in research upstream process development and cell culture manufacturing processes DMEM high glucose medium efficacy and homogeneity is tested DMEM media contain increased concentrations of vitamins minerals and amino acids compared to traditional basal media DMEM medium composition also includes ferric nitrate making this cell culture medium an option for certain cell lines requiring iron for cell growth DMEM media preparations are available in high or low glucose formulations as well as with or without L-glutamine magnesium and sodium pyruvate DMEM formulation options may also include phenol red as a pH indicator All available DMEM media products contain a sodium bicarbonate buffer with or without HEPES to maintain optimal culture pH DMEM raw material components are screened through strict quality control testing for lot-to-lot consistency DMEM liquid media products are hydrated using purified water and undergo sterile filtrat