Learn More
Alkali Scientific CellPro™ DMEM/Low glucose with L-glutamine, sodium pyruvate, 500ml. 10/Case
SDP
Catalog No. 503027200
Encompass_Preferred
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 50-302-7200 Supplier Alkali Scientific Supplier No. DMEM16CS
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.