Learn More
Alkali Scientific CellPro™ Donor Horse Serum, U.S. Origin, 500mL
SDP
Catalog No. 503026977
Encompass_Preferred
Donor horse serum can be used independently of other sera or as a supplement provided to enhance growth in combination with bovine sera. It is an ideal supplement to enhance growth of tumor cell lines, particularly of human origin, in conjunction with fetal bovine serum. It can also be used as a supplement in bacteriological media for growth enhancement of fastidious organisms. Donor horse serum is obtained from a closed, controlled standing herd that is under veterinary supervision and routinely monitored for various equine diseases. Sterile-filtered Mycoplasma-tested and virus-screened Coggins-tested for equine infectious anemia (swamp fever) Ships on dry ice. Store frozen at -20 to -10°C
Catalog No. 50-302-6977 Supplier Alkali Scientific Supplier No. FB32
Donor horse serum can be used independently of other sera or as a supplement provided to enhance growth in combination with bovine sera. It is an ideal supplement to enhance growth of tumor cell lines, particularly of human origin, in conjunction with fetal bovine serum. It can also be used as a supplement in bacteriological media for growth enhancement of fastidious organisms. Donor horse serum is obtained from a closed, controlled standing herd that is under veterinary supervision and routinely monitored for various equine diseases. Sterile-filtered Mycoplasma-tested and virus-screened Coggins-tested for equine infectious anemia (swamp fever) Ships on dry ice. Store frozen at -20 to -10°C