Learn More
Alkali Scientific CellPro™ DPBS 10X, 500ml, 10/Case
SDP
Catalog No. 503029107
Encompass_Preferred
Balanced salt solution used for multiple cell culture applications, (liquid form). Certificate of Analysis available electronically. Standard stock items from 100 mL to 200 L Volumes up to 1000 L available as made to order Not for diagnostic or therapeutic use. For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintenance of cell integrity is required for further manufacturing or research use.
Catalog No. 50-302-9107 Supplier Alkali Scientific Supplier No. PBD555CS
Balanced salt solution used for multiple cell culture applications, (liquid form). Certificate of Analysis available electronically. Standard stock items from 100 mL to 200 L Volumes up to 1000 L available as made to order Not for diagnostic or therapeutic use. For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintenance of cell integrity is required for further manufacturing or research use.