Learn More
Alkali Scientific CellPro™ DPBS Buffer, 1X, with Calcium and Magnesium, 1L
SDP
Supplier:  Alkali Scientific PBD1001
Encompass_Preferred
Alkali Scientific™ offers CellPro™ 1X DPBS, Dulbecco’s Phosphate Buffered Saline, a premium-grade solution meticulously formulated to meet the stringent requirements of cell culture and molecular biology applications. This balanced and isotonic buffer provides the ideal environment for cell maintenance, washing, and experimental procedures, ensuring optimal cell viability and functionality. Whether used for cell culture, immunocytochemistry, or protein assays, CellPro™ 1X DPBS delivers consistent and reliable results, making it an indispensable tool in biological research laboratories.
Catalog No. 50-238-9515