Learn More
Alkali Scientific CellPro™ DPBS Buffer, 1X, with Calcium and Magnesium, 500mL, 10/Case
SDP
Catalog No. 503028980
Encompass_Preferred
Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.
Catalog No. 50-302-8980 Supplier Alkali Scientific Supplier No. TBD555CS
Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.