Learn More
Alkali Scientific CellPro™ DPBS, Powder, Without Calcium & Magnesium, 10L
SDP
Catalog No. 5000662396
Encompass_Preferred
CellPro™ DPBS (Dulbecco’s Phosphate Buffered Saline) is a balanced salt solution in convenient powder form, designed for a wide range of cell culture and bioprocessing applications. Free of calcium and magnesium, this formulation is ideal for procedures requiring cell dissociation, rinsing chelators, or preparing cells for enzymatic treatments. • Balanced salt solution for maintaining osmotic balance • Calcium and magnesium free to prevent interference with cell dissociation enzymes • Pre weighed powder for fast and accurate 1X solution preparation • Suitable for cell washing, dilution, reagent reconstitution, and tissue transport • Manufactured in ISO 9001 certified, cGMP compliant facilities (21 CFR Part 820) • High purity raw materials ensure reproducible performance For research and further manufacturing use only. Not for diagnostic or therapeutic applications.
Catalog No. 50-006-62396 Supplier Alkali Scientific Supplier No. DPBS10
CellPro™ DPBS (Dulbecco’s Phosphate Buffered Saline) is a balanced salt solution in convenient powder form, designed for a wide range of cell culture and bioprocessing applications. Free of calcium and magnesium, this formulation is ideal for procedures requiring cell dissociation, rinsing chelators, or preparing cells for enzymatic treatments. • Balanced salt solution for maintaining osmotic balance • Calcium and magnesium free to prevent interference with cell dissociation enzymes • Pre weighed powder for fast and accurate 1X solution preparation • Suitable for cell washing, dilution, reagent reconstitution, and tissue transport • Manufactured in ISO 9001 certified, cGMP compliant facilities (21 CFR Part 820) • High purity raw materials ensure reproducible performance For research and further manufacturing use only. Not for diagnostic or therapeutic applications.