Learn More
Alkali Scientific CellPro™ DuoClick™ Two Position Screw-Cap Culture Tubes, 14mL/PS, 10/Racked, 500 /Case
SDP
Catalog No. 503029351
Encompass_Preferred
• Patented two-position screw-cap! • Aerobic culturing at first click • Anaerobic after second click • Printed graduations • Flat, markable screw-cap • Available racked DuoClick™ tubes feature a unique, patented screw closure system. DuoClick™ tubes completely eliminate the excess force, splashing and general frustration of traditional snap-cap culture tubes. The advanced cap design includes two distinct positions for aerobic and anaerobic culturing (see image no. 2). Just rotate the cap to the first click to secure the cap in the aerobic position. Or, rotate it past the second click for a leak-proof, airtight seal for anaerobic culturing. DuoClick™ tubes are available e-beam sterilized, with screw caps attached, and in our exclusive ready-to-use rack packaging. Polypropylene (PP) tubes can be centrifuged at up to 4,000×g and polystyrene (PS) tubes up to 1,400×g
Catalog No. 50-302-9351 Supplier Alkali Scientific Supplier No. CT8842
• Patented two-position screw-cap! • Aerobic culturing at first click • Anaerobic after second click • Printed graduations • Flat, markable screw-cap • Available racked DuoClick™ tubes feature a unique, patented screw closure system. DuoClick™ tubes completely eliminate the excess force, splashing and general frustration of traditional snap-cap culture tubes. The advanced cap design includes two distinct positions for aerobic and anaerobic culturing (see image no. 2). Just rotate the cap to the first click to secure the cap in the aerobic position. Or, rotate it past the second click for a leak-proof, airtight seal for anaerobic culturing. DuoClick™ tubes are available e-beam sterilized, with screw caps attached, and in our exclusive ready-to-use rack packaging. Polypropylene (PP) tubes can be centrifuged at up to 4,000×g and polystyrene (PS) tubes up to 1,400×g