Learn More
Alkali Scientific CellPro™ EGTA. 0.5M, pH8.0, 100mL
SDP
Catalog No. 502389397
Encompass_Preferred
Alkali Scientific™ CellPro™ EGTA, Molecular Biology Grade, is a meticulously purified compound designed to meet the stringent requirements of molecular biology and biochemistry applications. Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetra acetic acid (EGTA) is a chelating agent widely used for its ability to bind divalent cations, particularly calcium ions, effectively. This high-quality EGTA is meticulously prepared to ensure minimal impurities, making it suitable for critical molecular biology experiments such as nucleic acid and protein studies. With its exceptional purity and consistency, CellPro™ EGTA guarantees reliable and reproducible results in a wide range of research applications. Trust in the quality and performance of CellPro™ EGTA to support your molecular biology endeavors with precision and confidence.
Catalog No. 50-238-9397 Supplier Alkali Scientific Supplier No. DB0234
Alkali Scientific™ CellPro™ EGTA, Molecular Biology Grade, is a meticulously purified compound designed to meet the stringent requirements of molecular biology and biochemistry applications. Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetra acetic acid (EGTA) is a chelating agent widely used for its ability to bind divalent cations, particularly calcium ions, effectively. This high-quality EGTA is meticulously prepared to ensure minimal impurities, making it suitable for critical molecular biology experiments such as nucleic acid and protein studies. With its exceptional purity and consistency, CellPro™ EGTA guarantees reliable and reproducible results in a wide range of research applications. Trust in the quality and performance of CellPro™ EGTA to support your molecular biology endeavors with precision and confidence.