Learn More
Alkali Scientific CellPro™ Erlenmeyer Shake Flask, PC Conical with Baffles, Sterile, 1000mL, 6/Cs, Plug Seal Cap
SDP
Catalog No. 503027103
Encompass_Preferred
Erlenmeyer Baffled Flasks for shaker, storage, and titration functions are available. Choose flat bottom plastic or borosilicate glass options with durable white enamel scales or reinforced rims. Select disposable or autoclavable models with baffled bases in a variety of graduation ranges and capacities. Some flasks include both vented and solid screw caps for easy exchange. Sterile, individually wrapped, and pyrogen, DNase, and RNase free models are offered for cell culture applications. Heavy duty rim flasks with narrow neck and mouth from various suppliers are also available. Erlenmeyer flasks are sold in multipacks and economical case PC bottle Sterilized by E-Beam Nonpyrogenic Individually wrapped in a plastic bag. Both vent caps and plug seal caps are available. Vent caps with 0.22um hydrophobic filters for gas exchange without contamination.
Catalog No. 50-302-7103 Supplier Alkali Scientific Supplier No. EFB316
Erlenmeyer Baffled Flasks for shaker, storage, and titration functions are available. Choose flat bottom plastic or borosilicate glass options with durable white enamel scales or reinforced rims. Select disposable or autoclavable models with baffled bases in a variety of graduation ranges and capacities. Some flasks include both vented and solid screw caps for easy exchange. Sterile, individually wrapped, and pyrogen, DNase, and RNase free models are offered for cell culture applications. Heavy duty rim flasks with narrow neck and mouth from various suppliers are also available. Erlenmeyer flasks are sold in multipacks and economical case PC bottle Sterilized by E-Beam Nonpyrogenic Individually wrapped in a plastic bag. Both vent caps and plug seal caps are available. Vent caps with 0.22um hydrophobic filters for gas exchange without contamination.