Learn More
Alkali Scientific CellPro™ EXPRESS Vacuum Filtration Flasks PES Membrane, 12/Case, 0.22µm, 1000mL
SDP
Catalog No. 5000663279
Encompass_Preferred
CellPro™ EXPRESS Vacuum Filtration Flasks are designed for high speed and efficient filtration of tissue culture media, buffers, and other aqueous laboratory solutions. The system incorporates a low protein binding polyethersulfone (PES) membrane that enables filtration speeds up to 47% faster than standard filtration units, improving productivity in busy laboratory environments. Available in multiple volumes and membrane pore s, the system supports both small and large volume filtration workflows. Each unit includes a screw on funnel with lid, a graduated filtration nozzle compatible with standard vacuum hoses, and an ergonomically designed receiver bottle with grip indentations for easier handling. All components are gamma sterilized and certified RNase , DNase , and pyrogen free, ensuring safe and reliable filtration for sensitive biological samples.
Catalog No. 50-006-63279 Supplier Alkali Scientific Supplier No. V100022EXP
CellPro™ EXPRESS Vacuum Filtration Flasks are designed for high speed and efficient filtration of tissue culture media, buffers, and other aqueous laboratory solutions. The system incorporates a low protein binding polyethersulfone (PES) membrane that enables filtration speeds up to 47% faster than standard filtration units, improving productivity in busy laboratory environments. Available in multiple volumes and membrane pore s, the system supports both small and large volume filtration workflows. Each unit includes a screw on funnel with lid, a graduated filtration nozzle compatible with standard vacuum hoses, and an ergonomically designed receiver bottle with grip indentations for easier handling. All components are gamma sterilized and certified RNase , DNase , and pyrogen free, ensuring safe and reliable filtration for sensitive biological samples.