Learn More
Alkali Scientific CellPro™ Flow Cytometry Tubes with 35µm Mesh Strainer Cap, Sterile, 12 X 75mm, 25/Bag, 500/cs
SDP
Supplier:  Alkali Scientific CT6405
Encompass
Made of crystal clear polystyrene, these 12x75mm FlowTubes are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments. Designed for flow cytometry applications.
Catalog No. 50-233-6003