Learn More
Alkali Scientific CellPro™ Flow Cytometry Tubes without Cap, Sterile, 12 X 75mm, 250 per Tray Box, 1000/cs
SDP
Catalog No. 502335738
Encompass_Preferred
Made of crystal clear polystyrene, these 12x75mm FlowTubes are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments. Designed for flow cytometry applications.
Catalog No. 50-233-5738 Supplier Alkali Scientific Supplier No. CT6414
Made of crystal clear polystyrene, these 12x75mm FlowTubes are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences. The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments. Designed for flow cytometry applications.