Learn More
Alkali Scientific CellPro™ Flow Cytometry Cap Only with 70µm mesh, dual position, Opaque White, 500/Case
SDP
Catalog No. 503029211
Encompass_Preferred
We are pleased to announce the introduction of several new FACS / cell strainer caps for use with 5mL (12×75mm) culture tubes and FACS tubes. These cell strainers are ideal for detaching, isolating and straining cells from clusters and tissues. Their built in compatibility with 12×75mm culture tubes and FACS tubes makes them especially useful for pre-sorting and preparing samples for Flourescence-activated Cell Sorting (FACS). Typically a 35µm porous mesh is used to obtain a single-cell suspension and to prevent clogging in FACS instrumentation. However, a multitude of FACS users have cited applications requiring finer pore sizes for a higher level of anti-clog protection, while others require a larger pore size for pre-sorting and/or for more efficient rapid flow. • Pore sizes: 20µm, 35µm, 70µm, 100µm• Color coded by pore size• Sterile pouches of 25 strainer caps• E-Beam sterilized (SAL 10-6)
Catalog No. 50-302-9211 Supplier Alkali Scientific Supplier No. CT640970
We are pleased to announce the introduction of several new FACS / cell strainer caps for use with 5mL (12×75mm) culture tubes and FACS tubes. These cell strainers are ideal for detaching, isolating and straining cells from clusters and tissues. Their built in compatibility with 12×75mm culture tubes and FACS tubes makes them especially useful for pre-sorting and preparing samples for Flourescence-activated Cell Sorting (FACS). Typically a 35µm porous mesh is used to obtain a single-cell suspension and to prevent clogging in FACS instrumentation. However, a multitude of FACS users have cited applications requiring finer pore sizes for a higher level of anti-clog protection, while others require a larger pore size for pre-sorting and/or for more efficient rapid flow. • Pore sizes: 20µm, 35µm, 70µm, 100µm• Color coded by pore size• Sterile pouches of 25 strainer caps• E-Beam sterilized (SAL 10-6)