Learn More
Alkali Scientific CellPro™ Flow Cytometry Tubes, Compatible with FACS (BD) Instruments, Sterile, Individually wrapped, 500/Case
SDP
Catalog No. 503028595
Encompass_Preferred
CellPro™ Flow Cytometry Tubes are made of clear polystyrene and are designed for Fluorescence Activated Cell Sorting/flow cytometry equipment including FACS systems from Becton Dickinson (BD Biosciences). The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with automated instruments. Compatible with FACS (BD) instruments Available with 35µm strainer cap, standard cap, or no cap Clear polystyrene 5mL capacity Designed for flow cytometry/FACS applications
Catalog No. 50-302-8595 Supplier Alkali Scientific Supplier No. CT64051S
CellPro™ Flow Cytometry Tubes are made of clear polystyrene and are designed for Fluorescence Activated Cell Sorting/flow cytometry equipment including FACS systems from Becton Dickinson (BD Biosciences). The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with automated instruments. Compatible with FACS (BD) instruments Available with 35µm strainer cap, standard cap, or no cap Clear polystyrene 5mL capacity Designed for flow cytometry/FACS applications