Learn More
Alkali Scientific CellPro™ Fluid Thioglycollate Media 100mL
SDP
Catalog No. 502389037
Encompass_Preferred
Alkali Scientific™ CellPro™ Thioglycolate is a specialized medium designed for the cultivation of anaerobic and microaerophilic bacteria. With its unique formulation containing thioglycolate, dextrose, and other nutrients, this medium promotes bacterial growth under low-oxygen conditions. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for microbiological studies and microbial diversity analysis.
Catalog No. 50-238-9037 Supplier Alkali Scientific Supplier No. DC0009
Alkali Scientific™ CellPro™ Thioglycolate is a specialized medium designed for the cultivation of anaerobic and microaerophilic bacteria. With its unique formulation containing thioglycolate, dextrose, and other nutrients, this medium promotes bacterial growth under low-oxygen conditions. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for microbiological studies and microbial diversity analysis.