Learn More
Alkali Scientific CellPro™ Freezer box with hinged lid, 3", with insert for 25 snap-cap 5mL MacroTubes
SDP
Catalog No. 503027311
Encompass_Preferred
Alkali Scientific's 5mL MacroTube Cardboard Freezer Boxes are made of recyclable material, with a liquid-resistant outer coating. They are shipped fully assembled and are available in a 5×5 version that holds 25 x 5mL Snap-Cap tubes or a 6×6 version that holds 36 X 5mL Screw-Cap tubes. Each box has liquid nitrogen drainage slots and numbered grid markings in almost every conceivable location (top, bottom, sides, etc.).   Attached hinged lid 5×5 version for 5mL Snap-Caps 6×6 version for 5mL Screw-Caps Alpha-numeric coding Polycarbonate box fits 25 x 5mL tubes Polycarbonate box usable temp range is -196° to +121° Size: 5.25×5.25×3"  
Catalog No. 50-302-7311 Supplier Alkali Scientific Supplier No. DP443
Alkali Scientific's 5mL MacroTube Cardboard Freezer Boxes are made of recyclable material, with a liquid-resistant outer coating. They are shipped fully assembled and are available in a 5×5 version that holds 25 x 5mL Snap-Cap tubes or a 6×6 version that holds 36 X 5mL Screw-Cap tubes. Each box has liquid nitrogen drainage slots and numbered grid markings in almost every conceivable location (top, bottom, sides, etc.).   Attached hinged lid 5×5 version for 5mL Snap-Caps 6×6 version for 5mL Screw-Caps Alpha-numeric coding Polycarbonate box fits 25 x 5mL tubes Polycarbonate box usable temp range is -196° to +121° Size: 5.25×5.25×3"  