Learn More
Alkali Scientific CellPro™ Gentamicin Solution, 50 mg/mL, 10 x 10 mL
SDP
Catalog No. 503029282
Encompass_Preferred
An antibacterial and antimycobacterial reagent. Broad spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial levels Active against gram positive and gram-negative bacteria and mycoplasma; useful for long-term virus and tissue culture studies Interferes with bacterial protein synthesis by binding to L6 protein of 50S ribosomal subunit. Common working concentration is 5 μg/mL Used as an antibacterial or antimycobacterial.
Catalog No. 50-302-9282 Supplier Alkali Scientific Supplier No. FB93CS
An antibacterial and antimycobacterial reagent. Broad spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial levels Active against gram positive and gram-negative bacteria and mycoplasma; useful for long-term virus and tissue culture studies Interferes with bacterial protein synthesis by binding to L6 protein of 50S ribosomal subunit. Common working concentration is 5 μg/mL Used as an antibacterial or antimycobacterial.