Learn More
Alkali Scientific CellPro™ Ham's F12 medium, Kaighn's modification, 500ml
SDP
Catalog No. 503028832
Encompass_Preferred
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 50-302-8832 Supplier Alkali Scientific Supplier No. F12003
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.