Learn More
Alkali Scientific CellPro™ Ham's Nutrient Mixture F10 with L-glutamine, 500ml, 10/Case
SDP
Catalog No. 503027493
Encompass_Preferred
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 50-302-7493 Supplier Alkali Scientific Supplier No. F10001CS
Supports cell growth in research and biomanufacturing cell culture processes. Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.