Learn More
Alkali Scientific CellPro™ HBSS without calcium and magnesium, 500ml
SDP
Catalog No. 503027248
Encompass_Preferred
Balanced salt solution for cell culture applications (liquid form). All formulations incorporate a rigorous specification and approval process. Products with the LS and FS extension are supplied in convenient HyQPAK six-packs. Buffer cell culture growth with CellPro™s 1X Balanced Salt Solutions, available in lot sizes that facilitate scale-up and manufacturing.
Catalog No. 50-302-7248 Supplier Alkali Scientific Supplier No. HBSS03
Balanced salt solution for cell culture applications (liquid form). All formulations incorporate a rigorous specification and approval process. Products with the LS and FS extension are supplied in convenient HyQPAK six-packs. Buffer cell culture growth with CellPro™s 1X Balanced Salt Solutions, available in lot sizes that facilitate scale-up and manufacturing.