Learn More
Alkali Scientific CellPro™ IMDM modified medium with L-glutamine, HEPES, 10/Case
SDP
Catalog No. 503027441
Encompass_Preferred
Supports cell growth in research and biomanufacturing cell culture processes. Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.
Catalog No. 50-302-7441 Supplier Alkali Scientific Supplier No. IMDM01CS
Supports cell growth in research and biomanufacturing cell culture processes. Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities Raw material components screened through strict quality control testing for lot-to-lot consistency. Product efficacy and homogeneity tested. Products are hydrated using purified process water and undergo sterile filtration.