Learn More
Alkali Scientific CellPro Inoculating Loops Flex
SDP
Catalog No. 503028798
Encompass_Preferred
These sterile disposable plastic loops and needles are designed for use in quantitative procedures such as sampling urine counts and serial dilutions as well as for bacterial inoculation Because they are pre-sterilized and designed for single use there is no risk of cross-contamination and no need for flaming The unique hexagonal shape of the shaft makes it easier to grip orientate and guide through a smooth streaking process The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head This results in the elimination of cutting or gouging of the agar surface The flexibility of polypropylene loops makes it easier to maneuver them within Petri dishes test tubes or other confined spaces improving the researchers ability to collect and transfer samples efficiently The flexibility allows for more precise sampling from cultures especially when working with small or delicate colonies that could be easily damaged by more rigid instruments
Catalog No. 50-302-8798 Supplier Alkali Scientific Supplier No. M2025W
These sterile disposable plastic loops and needles are designed for use in quantitative procedures such as sampling urine counts and serial dilutions as well as for bacterial inoculation Because they are pre-sterilized and designed for single use there is no risk of cross-contamination and no need for flaming The unique hexagonal shape of the shaft makes it easier to grip orientate and guide through a smooth streaking process The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head This results in the elimination of cutting or gouging of the agar surface The flexibility of polypropylene loops makes it easier to maneuver them within Petri dishes test tubes or other confined spaces improving the researchers ability to collect and transfer samples efficiently The flexibility allows for more precise sampling from cultures especially when working with small or delicate colonies that could be easily damaged by more rigid instruments