Learn More
Alkali Scientific ASI™ CellPro™ Internally Threaded Cryovials, 0.5mL, Sterile, 500/cs
SDP
Catalog No. 503028162
Encompass_Preferred
CellPro™ Cryovials are designed for safe storage of samples at temperatures as low as -196 °C (gas phase of liquid nitrogen only). Silicone O-Rings on internally threaded caps create a leakproof seal. All CellPro Croyvials are sterilized by E-Beam and are Non-pyrogenic, DNase/RNase and human DNA free. Sterilized by E-Beam Ultra Low Temp Non-pyrogenic, DNase/RNase and human DNA free Internally  threaded caps Round bottom, self-standing Recommend for storage in gas phase of nitrogen tank
Catalog No. 50-302-8162 Supplier Alkali Scientific Supplier No. VCI08
CellPro™ Cryovials are designed for safe storage of samples at temperatures as low as -196 °C (gas phase of liquid nitrogen only). Silicone O-Rings on internally threaded caps create a leakproof seal. All CellPro Croyvials are sterilized by E-Beam and are Non-pyrogenic, DNase/RNase and human DNA free. Sterilized by E-Beam Ultra Low Temp Non-pyrogenic, DNase/RNase and human DNA free Internally  threaded caps Round bottom, self-standing Recommend for storage in gas phase of nitrogen tank