Learn More
Alkali Scientific CellPro™ IPTG Solution 5mM 10mL
SDP
Catalog No. 502389125
Encompass_Preferred
CellPro™ IPTG (Isopropyl β-D-1-thiogalactopyranoside) Solution stands as a beacon of quality and reliability in molecular biology research. Crafted to meticulous standards of purity and sterility, this solution provides researchers with a reliable inducer for gene expression studies. With its sterile filtered formulation and concentration of 5mM in a 10mL volume, CellPro™ IPTG Solution ensures precise and reproducible results, empowering researchers in their molecular biology experiments.
Catalog No. 50-238-9125 Supplier Alkali Scientific Supplier No. DS0176
Add to Cart
Add to Cart
CellPro™ IPTG (Isopropyl β-D-1-thiogalactopyranoside) Solution stands as a beacon of quality and reliability in molecular biology research. Crafted to meticulous standards of purity and sterility, this solution provides researchers with a reliable inducer for gene expression studies. With its sterile filtered formulation and concentration of 5mM in a 10mL volume, CellPro™ IPTG Solution ensures precise and reproducible results, empowering researchers in their molecular biology experiments.