Learn More
Alkali Scientific CellPro™ L-15 Leibovitz media with L-glutamine, 500ml
SDP
Catalog No. 503027655
Encompass_Preferred
Supports cell growth in research and biomanufacturing cell culture processes. Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities. Raw material components screened through strict quality control testing for lot-to-lot consistency. Products are hydrated using purified process water and undergo sterile filtration
Catalog No. 50-302-7655 Supplier Alkali Scientific Supplier No. L1501
Supports cell growth in research and biomanufacturing cell culture processes. Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities. Raw material components screened through strict quality control testing for lot-to-lot consistency. Products are hydrated using purified process water and undergo sterile filtration