Learn More
Alkali Scientific CellPro™ LB + Ampicillin 50µg/mL + Tetracycline 15µg/mL, 10/Sleeve
SDP
Catalog No. 502389505
Encompass_Preferred
CellPro™ LB + Ampicillin is a specialized bacterial growth medium designed for the cultivation of ampicillin-resistant bacterial strains. This nutrient-rich agar contains ampicillin, an antibiotic that inhibits the growth of bacteria lacking ampicillin resistance genes. Ideal for molecular biology and microbiology applications, this medium promotes the selective growth of genetically modified bacteria harboring ampicillin resistance plasmids, facilitating the isolation and maintenance of recombinant DNA constructs.
Catalog No. 50-238-9505 Supplier Alkali Scientific Supplier No. DC0052
CellPro™ LB + Ampicillin is a specialized bacterial growth medium designed for the cultivation of ampicillin-resistant bacterial strains. This nutrient-rich agar contains ampicillin, an antibiotic that inhibits the growth of bacteria lacking ampicillin resistance genes. Ideal for molecular biology and microbiology applications, this medium promotes the selective growth of genetically modified bacteria harboring ampicillin resistance plasmids, facilitating the isolation and maintenance of recombinant DNA constructs.