Learn More
Alkali Scientific CellPro™ LB Broth 100mL
SDP
Catalog No. 502389122
Encompass_Preferred
Alkali Scientific™ CellPro™ LB Broth is a premium-quality growth medium formulated for the cultivation of Escherichia coli and other bacterial strains in molecular biology applications. With its precise composition of tryptone, yeast extract, and sodium chloride, this broth supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ LB Broth a reliable choice for bacterial culture, plasmid DNA isolation, and recombinant protein production.
Catalog No. 50-238-9122 Supplier Alkali Scientific Supplier No. DC0007
Alkali Scientific™ CellPro™ LB Broth is a premium-quality growth medium formulated for the cultivation of Escherichia coli and other bacterial strains in molecular biology applications. With its precise composition of tryptone, yeast extract, and sodium chloride, this broth supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ LB Broth a reliable choice for bacterial culture, plasmid DNA isolation, and recombinant protein production.