Learn More
Alkali Scientific CellPro™ LB Broth + Magnesium Sulfate 500mL
SDP
Catalog No. 502388811
Encompass_Preferred
Alkali Scientific™ CellPro™ LB Broth is a premium-quality growth medium formulated for the cultivation of Escherichia coli and other bacterial strains in molecular biology applications. With its precise composition of tryptone, yeast extract, and sodium chloride, this broth supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ LB Broth a reliable choice for bacterial culture, plasmid DNA isolation, and recombinant protein production.
Catalog No. 50-238-8811 Supplier Alkali Scientific Supplier No. DC0018
Add to Cart
Add to Cart
Alkali Scientific™ CellPro™ LB Broth is a premium-quality growth medium formulated for the cultivation of Escherichia coli and other bacterial strains in molecular biology applications. With its precise composition of tryptone, yeast extract, and sodium chloride, this broth supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ LB Broth a reliable choice for bacterial culture, plasmid DNA isolation, and recombinant protein production.