Learn More
Alkali Scientific CellPro™ LB + Carbenicillin 50µg/mL + Tetracycline 12.5µg/mL + Xgal 40µg/mL, 10/Sleeve
SDP
Catalog No. 502389103
Encompass_Preferred
CellPro™ LB + Carbenicillin is a specialized culture medium containing carbenicillin, a β-lactam antibiotic used to select for bacterial strains carrying carbenicillin resistance genes. This LB agar variant is commonly employed in molecular biology research for the cultivation and maintenance of recombinant bacterial cells harboring carbenicillin resistance plasmids. By incorporating carbenicillin into the LB agar, this medium enables the selective growth of transformed bacterial strains while inhibiting the growth of non-resistant contaminants.
Catalog No. 50-238-9103 Supplier Alkali Scientific Supplier No. DC0084
Add to Cart
Add to Cart
CellPro™ LB + Carbenicillin is a specialized culture medium containing carbenicillin, a β-lactam antibiotic used to select for bacterial strains carrying carbenicillin resistance genes. This LB agar variant is commonly employed in molecular biology research for the cultivation and maintenance of recombinant bacterial cells harboring carbenicillin resistance plasmids. By incorporating carbenicillin into the LB agar, this medium enables the selective growth of transformed bacterial strains while inhibiting the growth of non-resistant contaminants.