Learn More
Alkali Scientific CellPro™ LB (Lennox) + Kanamycin 50µg/mL + Tetracycline 25µg/mL 100mm, 10/Sleeve
SDP
Catalog No. 502389126
Encompass_Preferred
Alkali Scientific™ CellPro™ LB (Lennox) is a molecular biology-grade growth medium designed for the cultivation of Escherichia coli strains. With its balanced composition of tryptone, yeast extract, and sodium chloride, this media supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to maintain purity and consistency, ensuring optimal performance in bacterial culture, plasmid DNA isolation, and recombinant protein production.
Catalog No. 50-238-9126 Supplier Alkali Scientific Supplier No. DC0119
Alkali Scientific™ CellPro™ LB (Lennox) is a molecular biology-grade growth medium designed for the cultivation of Escherichia coli strains. With its balanced composition of tryptone, yeast extract, and sodium chloride, this media supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to maintain purity and consistency, ensuring optimal performance in bacterial culture, plasmid DNA isolation, and recombinant protein production.