Learn More
Alkali Scientific CellPro™ LB + Kanamycin 50µg/mL + Xgal 40µg/mL + IPTG .5mM 100mm, 10/Sleeve
SDP
Catalog No. 502388770
Encompass_Preferred
Alkali Scientific™ CellPro™ LB is a molecular biology-grade growth medium designed for the cultivation of Escherichia coli strains. With its balanced composition of tryptone, yeast extract, and sodium chloride, this media supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to maintain purity and consistency, ensuring optimal performance in bacterial culture, plasmid DNA isolation, and recombinant protein production.
Catalog No. 50-238-8770 Supplier Alkali Scientific Supplier No. DC0121
Alkali Scientific™ CellPro™ LB is a molecular biology-grade growth medium designed for the cultivation of Escherichia coli strains. With its balanced composition of tryptone, yeast extract, and sodium chloride, this media supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to maintain purity and consistency, ensuring optimal performance in bacterial culture, plasmid DNA isolation, and recombinant protein production.