Learn More
Alkali Scientific CellPro™ LB (Lennox) + Ampicillin 100µg/mL + Kanamycin 30µg/mL, 10/Sleeve
SDP
Catalog No. 502388734
Encompass_Preferred
CellPro™ LB (Lennox) + Ampicillin is a modified variant of the classic Luria-Bertani (LB) agar, fortified with ampicillin to support the growth of ampicillin-resistant bacterial strains. This medium is commonly used in molecular biology laboratories for the cultivation and selection of transformed bacterial cells carrying ampicillin resistance genes. By incorporating ampicillin into the LB agar, this medium enables the isolation and propagation of genetically engineered bacteria in a controlled and selective manner.
Catalog No. 50-238-8734 Supplier Alkali Scientific Supplier No. DC0079
CellPro™ LB (Lennox) + Ampicillin is a modified variant of the classic Luria-Bertani (LB) agar, fortified with ampicillin to support the growth of ampicillin-resistant bacterial strains. This medium is commonly used in molecular biology laboratories for the cultivation and selection of transformed bacterial cells carrying ampicillin resistance genes. By incorporating ampicillin into the LB agar, this medium enables the isolation and propagation of genetically engineered bacteria in a controlled and selective manner.