Learn More
Alkali Scientific CellPro™ LB (Lennox) + Carbenicillin 100µg/mL 100mm, 10/Sleeve
SDP
Supplier:  Alkali Scientific DC0090
Encompass_Preferred
CellPro™ LB Lennox Agar is a modification of Luria-Bertani (LB) agar, formulated with reduced sodium chloride content compared to standard LB agar. This medium is commonly used for molecular biology applications, including the cultivation and selection of recombinant bacterial strains. LB Lennox Agar provides a nutrient-rich environment for bacterial growth, supporting the propagation of transformed cells harboring plasmids with antibiotic resistance markers.
Catalog No. 50-238-8712