Learn More
Alkali Scientific CellPro™ LB (Lennox) + Chloramphenicol 50µg/mL 150mm
SDP
Catalog No. 502388783
Encompass_Preferred
CellPro™ LB (Lennox) + Chloramphenicol is a specialized culture medium formulated for the growth of bacterial strains resistant to chloramphenicol. This LB agar variant contains chloramphenicol, a bacteriostatic antibiotic that inhibits protein synthesis in susceptible bacteria. By incorporating chloramphenicol into the LB agar, this medium selectively promotes the growth of chloramphenicol-resistant bacterial strains, making it ideal for the maintenance of recombinant DNA constructs and other molecular biology applications.
Catalog No. 50-238-8783 Supplier Alkali Scientific Supplier No. DC0159
CellPro™ LB (Lennox) + Chloramphenicol is a specialized culture medium formulated for the growth of bacterial strains resistant to chloramphenicol. This LB agar variant contains chloramphenicol, a bacteriostatic antibiotic that inhibits protein synthesis in susceptible bacteria. By incorporating chloramphenicol into the LB agar, this medium selectively promotes the growth of chloramphenicol-resistant bacterial strains, making it ideal for the maintenance of recombinant DNA constructs and other molecular biology applications.