Learn More
Alkali Scientific CellPro™ LB Media + 1.5% Agar 500mL in 1L Bottle
SDP
Catalog No. 502389485
Encompass_Preferred
Alkali Scientific™ CellPro™ LB Media is a rich bacterial growth medium widely used in microbiology and molecular biology research. With its balanced composition of peptone, yeast extract, and sodium chloride, this media supports robust bacterial growth and protein expression. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for bacterial culture, protein production, and recombinant DNA technology.
Catalog No. 50-238-9485 Supplier Alkali Scientific Supplier No. DC0035
Add to Cart
Add to Cart
Alkali Scientific™ CellPro™ LB Media is a rich bacterial growth medium widely used in microbiology and molecular biology research. With its balanced composition of peptone, yeast extract, and sodium chloride, this media supports robust bacterial growth and protein expression. Each batch undergoes rigorous sterile filtration to maintain purity and consistency, providing researchers with a reliable tool for bacterial culture, protein production, and recombinant DNA technology.