Learn More
Alkali Scientific CellPro™ Low Evap 6-Well Cell Culture Plates, Flat Bottom, Sterile, 50/Case, Tissue Treated
SDP
Catalog No. 503028926
Encompass_Preferred
CellPro™ Cell Culture Plates have round edges for better handling. Low profile, low-evaporation lid design assure uniform growth. Tissue treated and non-treated are both available. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. Compare with common cell culture plate: grooves are designed at the edge of the culture plate to avoid the edge effect to the greatest extent, and to ensure that the cells maintain their optimal state during cell culture.
Catalog No. 50-302-8926 Supplier Alkali Scientific Supplier No. TPN1006TLEVP
CellPro™ Cell Culture Plates have round edges for better handling. Low profile, low-evaporation lid design assure uniform growth. Tissue treated and non-treated are both available. Non-treated growth surface is more hydrophobic than tissue culture surface and are similar to the surface of a bacteriological Petri dish. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. Compare with common cell culture plate: grooves are designed at the edge of the culture plate to avoid the edge effect to the greatest extent, and to ensure that the cells maintain their optimal state during cell culture.